This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00550635_m1
See other TCF25 GE Assays ›
Gene Symbol:
Tcf25
Gene Name:
transcription factor 25 (basic helix-loop-helix)
Gene Aliases:
1100001J13Rik, 1810041K11Rik, D8Ertd325e, Nulp1, mKIAA1049
Chromosome Location:
Chr.8: 123373635 - 123404174 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00550635_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tcf25
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037877.3 NP_001032966.1 4 - 5 709 70
NM_001037878.3 NP_001032967.1 4 - 5 709 70
NM_001286362.1 NP_001273291.1 3 - 4 634 70
NM_001286363.1 NP_001273292.1 3 - 4 634 70
NM_025804.3 NP_080080.2 4 - 5 709 70
GenBank mRNA AK003180.1 - 4 - 5 597 70
AK013754.1 - 4 - 5 398 70
AK045397.1 - 3 - 4 562 70
AK050274.1 - 4 - 5 637 70
AK075728.1 - 4 - 5 563 70
AK088360.1 - 4 - 5 597 70
AK089964.1 - 4 - 5 598 70
AK122430.1 - 4 - 5 564 70
AK148082.1 - 4 - 5 571 70
AK150232.1 - 4 - 5 596 70
AK150918.1 - 4 - 5 590 70
AK150942.1 - 4 - 5 583 70
AK151143.1 - 4 - 5 583 70
AK151558.1 - 4 - 5 590 70
AK152178.1 - 4 - 5 583 70
AK152494.1 - 4 - 5 583 70
AK161627.1 - 4 - 5 585 70
AK167837.1 - 4 - 5 583 70
AK168167.1 - 4 - 5 583 70
BC016130.1 - 4 - 5 571 70
BC025071.1 - 4 - 5 570 3498003 70
BC046768.1 - 4 - 5 667 5707312 70
EU624323.1 - 3 - 4 472 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001520.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK013754.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tcf25

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800012450.mm.1 NM_001037877.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012450.mm.1 NM_001037878.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012450.mm.1 NM_001286362.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800012450.mm.1 NM_001286363.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800012450.mm.1 NM_025804.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012450.mm.1 TC0800001520.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0