This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00550637_m1
See other TCF25 GE Assays ›
Gene Symbol:
Tcf25
Gene Name:
transcription factor 25 (basic helix-loop-helix)
Gene Aliases:
1100001J13Rik, 1810041K11Rik, D8Ertd325e, Nulp1, mKIAA1049
Chromosome Location:
Chr.8: 123373635 - 123404174 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00550637_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tcf25
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037877.3 NP_001032966.1 6 - 7 857 69
NM_001037878.3 NP_001032967.1 6 - 7 857 69
NM_001286362.1 NP_001273291.1 5 - 6 782 69
NM_001286363.1 NP_001273292.1 5 - 6 782 69
NM_025804.3 NP_080080.2 6 - 7 857 69
GenBank mRNA AK003180.1 - 6 - 7 745 69
AK013754.1 - 6 - 7 546 69
AK045397.1 - 5 - 6 710 69
AK050274.1 - 6 - 7 785 69
AK075728.1 - 6 - 7 711 69
AK088360.1 - 6 - 7 745 69
AK089964.1 - 6 - 7 746 69
AK122430.1 - 6 - 7 712 69
AK148082.1 - 6 - 7 719 69
AK150232.1 - 6 - 7 744 69
AK150918.1 - 6 - 7 738 69
AK150942.1 - 6 - 7 731 69
AK151143.1 - 6 - 7 731 69
AK151558.1 - 6 - 7 738 69
AK152178.1 - 6 - 7 731 69
AK152494.1 - 6 - 7 731 69
AK161627.1 - 6 - 7 733 69
AK198411.1 - 3 - 4 168 69
BC016130.1 - 6 - 7 719 69
BC025071.1 - 6 - 7 718 3498003 69
BC046768.1 - 6 - 7 815 5707312 69
EU624323.1 - 5 - 6 620 69
U94988.1 - 6 - 7 718 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001520.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK013754.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tcf25

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800012452.mm.1 NM_001037877.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012452.mm.1 NM_001037878.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012452.mm.1 NM_001286362.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800012452.mm.1 NM_001286363.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800012452.mm.1 NM_025804.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012452.mm.1 TC0800001520.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0