This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00550638_m1
See other TCF25 GE Assays ›
Gene Symbol:
Tcf25
Gene Name:
transcription factor 25 (basic helix-loop-helix)
Gene Aliases:
1100001J13Rik, 1810041K11Rik, D8Ertd325e, Nulp1, mKIAA1049
Chromosome Location:
Chr.8: 123373635 - 123404174 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00550638_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tcf25
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037877.3 NP_001032966.1 7 - 8 991 70
NM_001037878.3 NP_001032967.1 7 - 8 991 70
NM_001286362.1 NP_001273291.1 6 - 7 916 70
NM_001286363.1 NP_001273292.1 6 - 7 916 70
NM_025804.3 NP_080080.2 7 - 8 991 70
GenBank mRNA AK003180.1 - 7 - 8 879 70
AK013754.1 - 7 - 8 680 70
AK045397.1 - 6 - 7 844 70
AK050274.1 - 7 - 8 919 70
AK075728.1 - 7 - 8 845 70
AK076151.1 - 1 - 2 78 70
AK088360.1 - 7 - 8 879 70
AK089964.1 - 7 - 8 880 70
AK122430.1 - 7 - 8 846 70
AK148082.1 - 7 - 8 853 70
AK150232.1 - 7 - 8 878 70
AK150918.1 - 7 - 8 872 70
AK150942.1 - 7 - 8 865 70
AK151143.1 - 7 - 8 865 70
AK151558.1 - 7 - 8 872 70
AK152178.1 - 7 - 8 865 70
AK152494.1 - 7 - 8 865 70
AK161627.1 - 7 - 8 867 70
AK167837.1 - 7 - 8 865 70
AK168167.1 - 7 - 8 865 70
AK198411.1 - 4 - 5 302 70
AK210828.1 - 2 - 3 82 70
BC016130.1 - 7 - 8 853 70
BC025071.1 - 7 - 8 852 3498003 70
BC046768.1 - 7 - 8 949 5707312 70
EU624323.1 - 6 - 7 754 70
U94988.1 - 7 - 8 852 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001520.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK076151.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tcf25

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800012453.mm.1 NM_001037877.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012453.mm.1 NM_001037878.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012453.mm.1 NM_001286362.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800012453.mm.1 NM_001286363.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800012453.mm.1 NM_025804.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012453.mm.1 TC0800001520.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800012455.mm.1 NM_001037877.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012455.mm.1 NM_001037878.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012455.mm.1 NM_001286362.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800012455.mm.1 NM_001286363.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800012455.mm.1 NM_025804.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012455.mm.1 TC0800001520.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-kg5vd:80/100.66.78.64:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0