This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00550646_m1
See other TCF25 GE Assays ›
Gene Symbol:
Tcf25
Gene Name:
transcription factor 25 (basic helix-loop-helix)
Gene Aliases:
1100001J13Rik, 1810041K11Rik, D8Ertd325e, Nulp1, mKIAA1049
Chromosome Location:
Chr.8: 123373635 - 123404174 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00550646_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tcf25
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037877.3 NP_001032966.1 15 - 16 1887 66
NM_001037878.3 NP_001032967.1 15 - 16 1887 66
NM_001286362.1 NP_001273291.1 14 - 15 1812 66
NM_001286363.1 NP_001273292.1 14 - 15 1812 66
NM_025804.3 NP_080080.2 15 - 16 1887 66
GenBank mRNA AK003180.1 - 15 - 16 1774 66
AK013754.1 - 15 - 16 1576 66
AK045397.1 - 14 - 15 1740 66
AK050274.1 - 15 - 16 1813 66
AK075728.1 - 15 - 16 1737 66
AK076151.1 - 9 - 10 974 66
AK088360.1 - 15 - 16 1775 66
AK088602.1 - 2 - 3 426 66
AK089964.1 - 15 - 16 1776 66
AK122430.1 - 15 - 16 1742 66
AK148082.1 - 15 - 16 1749 66
AK150918.1 - 15 - 16 1768 66
AK150942.1 - 15 - 16 1761 66
AK151143.1 - 15 - 16 1761 66
AK151558.1 - 15 - 16 1767 66
AK152178.1 - 15 - 16 1761 66
AK152494.1 - 15 - 16 1761 66
AK161627.1 - 15 - 16 1763 66
AK167837.1 - 15 - 16 1761 66
AK168167.1 - 15 - 16 1761 66
AK191661.1 - 1 - 2 53 66
BC016130.1 - 15 - 16 1749 66
BC025071.1 - 15 - 16 1748 3498003 66
BC046768.1 - 15 - 16 1845 5707312 66
EU624323.1 - 14 - 15 1650 66
U94988.1 - 15 - 16 1748 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001520.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tcf25

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800012466.mm.1 NM_001037877.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012466.mm.1 NM_001037878.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012466.mm.1 NM_001286362.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800012466.mm.1 NM_001286363.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800012466.mm.1 NM_025804.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012466.mm.1 TC0800001520.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800012465.mm.1 NM_001037877.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012465.mm.1 NM_001037878.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012465.mm.1 NM_001286362.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800012465.mm.1 NM_001286363.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800012465.mm.1 NM_025804.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012465.mm.1 TC0800001520.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0