This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00550933_m1
See other GORASP2 GE Assays ›
Gene Symbol:
Gorasp2
Gene Name:
golgi reassembly stacking protein 2
Gene Aliases:
0610011A07Rik, 2410043M02Rik, 5730520M13Rik, 9430094F20Rik, AW552058, GOLPH2, GRASP55, GRS2, p59
Chromosome Location:
Chr.2: 70661489 - 70691725 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00550933_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gorasp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027352.4 NP_081628.3 5 - 6 751 71
NR_027343.1 6 - 7 948 71
GenBank mRNA AK002515.1 - 6 - 7 830 71
AK010664.2 - 5 - 6 670 71
AK033413.1 - 5 - 6 685 71
AK044691.1 - 636 71
AK077153.1 - 5 - 6 626 71
AK077683.1 - 5 - 6 686 71
AK077706.1 - 5 - 6 618 71
AK088239.1 - 5 - 6 632 71
AK148337.1 - 5 - 6 685 71
AK151843.1 - 5 - 6 633 71
AK152475.1 - 5 - 6 633 71
AK152544.1 - 5 - 6 633 71
AK152603.1 - 5 - 6 661 71
AK153049.1 - 5 - 6 633 71
AK153555.1 - 5 - 6 684 71
AK159469.1 - 5 - 6 683 71
AK167823.1 - 5 - 6 685 71
AK168928.1 - 6 - 7 881 71
AK169382.1 - 5 - 6 633 71
BC005600.1 - 5 - 6 624 3482776 71
BC016455.1 - 4 - 5 476 4234343 71
BC024982.1 - 5 - 6 637 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200001025.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gorasp2

Gene Ontology Categories:

Function(s) Process(es)

organelle organization
Golgi organization
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200008225.mm.1 NM_027352.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200008225.mm.1 NM_027352.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200008225.mm.1 NR_027343.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200008225.mm.1 NR_027343.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200008225.mm.1 TC0200001025.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-7669c5b596-8xbxd:80/100.66.76.149:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0