This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00550965_m1
See other RNF167 GE Assays ›
Gene Symbol:
Rnf167
Gene Name:
ring finger protein 167
Gene Aliases:
0610010G05Rik, 5730408C10Rik, AU019305, AV328608
Chromosome Location:
Chr.11: 70647589 - 70651421 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00550965_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rnf167
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027445.1 NP_081721.1 2 - 3 444 90
GenBank mRNA AK075579.1 - 2 - 3 473 90
AK154071.1 - 2 - 3 432 90
BC010777.1 - 2 - 3 444 4241357 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100000961.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rnf167

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
negative regulation of cell cycle
ubiquitin-protein transferase activity
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2012779 AK154071.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2012779 BC010777.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2012779 AK075579.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2012779 AK030908.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2012779 NM_027445.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2012780 AK154071.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2012780 BC010777.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2012780 AK045166.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2012780 AK075579.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2012780 NM_027445.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 524528 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 293926 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2012780 AK154071.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2012780 BC010777.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2012780 AK045166.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2012780 AK075579.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2012780 NM_027445.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2012779 AK154071.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2012779 BC010777.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2012779 AK075579.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2012779 AK030908.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2012779 NM_027445.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901852 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901838 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901861 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901849 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901837 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901847 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901843 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901863 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901839 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901853 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901858 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901866 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901864 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901841 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901840 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901845 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901835 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901868 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901855 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901850 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901844 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07389148 - -
Whole Mouse Genome G4122A A_52_P484281 AK154071.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P484281 BC010777.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P484281 AK075579.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P484281 NM_027445.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P484281 AK017523.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140609 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103528 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09578933 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09578932 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901904 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901888 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901901 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901907 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901915 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901945 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901844 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901941 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901895 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901847 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901911 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901863 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901917 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901885 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901838 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901892 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901850 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901794 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901908 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901930 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901835 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901866 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901953 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901940 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901875 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901832 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901936 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901914 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901853 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901846 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901886 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901841 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901873 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901877 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901855 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901849 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901833 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901858 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901920 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901947 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901869 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901899 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901942 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901926 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901910 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901868 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901924 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901830 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901918 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901837 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901922 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901897 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901861 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901950 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901879 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901864 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901938 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901898 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901929 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901891 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901840 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901843 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27901855 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27901843 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09707221 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27901841 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09707220 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27901832 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27901838 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27901868 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27901845 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27901835 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27901858 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27901864 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27901840 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27901844 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27901837 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27901866 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09707226 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27901863 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27901852 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09565432 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09707216 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09707215 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27901833 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10377933 - -
Affymetrix Mouse Gene 1.0 ST Array 10377929 - -
Affymetrix Mouse Gene 1.0 ST Array 10377935 - -
Affymetrix Mouse Gene 1.0 ST Array 10377932 - -
Affymetrix Mouse Gene 1.0 ST Array 10377930 - -
Affymetrix Mouse Gene 1.0 ST Array 10377931 - -
Affymetrix Mouse Gene 1.0 ST Array 10377936 - -
Affymetrix Mouse Gene 1.0 ST Array 10377928 - -
Affymetrix Mouse Gene 1.0 ST Array 10377937 - -
Affymetrix Mouse Gene 1.0 ST Array 10377934 - -
Affymetrix HT MG-430 PM Array Plate 1430527_PM_a_at - -
Affymetrix Mouse Gene 1.1 ST Array 10377927 - -
Mouse Genome 430 2.0 Array 1430527_a_at - -
Mouse Genome 430 2.0 Array 1430527_PM_a_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 70510_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 70510_at - -
Affymetrix Mouse Clariom S Array PSR1100008574.mm.1 NM_027445.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100008574.mm.1 TC1100000961.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100008581.mm.1 NM_027445.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100008581.mm.1 TC1100000961.mm.1 MTA TC
Mouse Expression Set 430 1430527_PM_a_at - -
Mouse Expression Set 430 1430527_a_at - -
Murine Genome U74v2 Set 94555_at - -
Murine Genome U74v2 Set 94555_PM_at - -
Mouse Genome 430A 2.0 Array 1430527_PM_a_at - -
Mouse Genome 430A 2.0 Array 1430527_a_at - -
Affymetrix HT Mouse Genome 430A Array 1430527_a_at - -
Affymetrix HT Mouse Genome 430A Array 1430527_PM_a_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 1500333 - -
Illumina mouse-6 v1.0 expression beadchip 1410026 - -
Illumina mouse-6 v1.0 expression beadchip 1500333 - -
Illumina mouse-6 v1.0 expression beadchip 2810091 - -
Illumina mouse-6 v1.0 expression beadchip 4730093 - -
Illumina mouseRef-8 v1.0 expression beadchip 0001500333 - -
Illumina mouseRef-8 v1.0 expression beadchip 0004730093 - -
Illumina mouseRef-8 v1.0 expression beadchip 0002810091 - -
Illumina mouseRef-8 v1.0 expression beadchip 0001410026 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1214800 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2687744 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2698372 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1229534 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1229534 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2687744 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2698372 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1214800 - -
mouse-6 v1.1 (Illumina) scl41328.5.1_14-S - -
mouse-6 v1.1 (Illumina) scl0001587.1_50-S - -
mouse-6 v1.1 (Illumina) scl0001478.1_0-S - -
mouse-6 v1.1 (Illumina) scl070510.9_205-S - -
Illumina Sentrix mouse-6.v1 bead array 1500333 - -
Illumina Sentrix mouse-6.v1 bead array 2810091 - -
Illumina Sentrix mouse-6.v1 bead array 4730093 - -
Illumina Sentrix mouse-6.v1 bead array 1410026 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0001587.1_50-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl41328.5.1_14-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0001478.1_0-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl070510.9_205-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 4730093 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 1500333 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 2810091 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 1410026 - -
MouseRef-8 v2 Expression BeadChip ILMN_2892860 - -
MouseRef-8 v2 Expression BeadChip ILMN_2687744 - -
MouseRef-8 v2 Expression BeadChip ILMN_1229534 - -
MouseRef-8 v2 Expression BeadChip ILMN_2892856 - -
MouseRef-8 v2 Expression BeadChip ILMN_1214800 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2687744 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2892856 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2892860 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1214800 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1229534 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl070510.9_205-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0001587.1_50-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl41328.5.1_14-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0001478.1_0-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-flftk:80/100.66.79.227:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0