This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00550966_g1
See other RNF167 GE Assays ›
Gene Symbol:
Rnf167
Gene Name:
ring finger protein 167
Gene Aliases:
0610010G05Rik, 5730408C10Rik, AU019305, AV328608
Chromosome Location:
Chr.11: 70647589 - 70651421 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00550966_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rnf167
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027445.1 NP_081721.1 3 - 4 572 76
GenBank mRNA AK075579.1 - 3 - 4 601 76
AK154071.1 - 3 - 4 560 76
BC010777.1 - 3 - 4 572 4241357 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100000961.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rnf167

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
negative regulation of cell cycle
ubiquitin-protein transferase activity
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100008582.mm.1 NM_027445.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100008582.mm.1 TC1100000961.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100008584.mm.1 NM_027445.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100008584.mm.1 TC1100000961.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0