This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00551149_g1
See other RPUSD1 GE Assays ›
Gene Symbol:
Rpusd1
Gene Name:
RNA pseudouridylate synthase domain containing 1
Gene Aliases:
2310051D06Rik, AU019540, Rlucl
Chromosome Location:
Chr.17: 25727703 - 25731464 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00551149_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rpusd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028009.3 NP_082285.1 1 - 2 98 68
GenBank mRNA AK049334.1 - 1 - 2 99 68
BC018186.1 - 1 - 2 59 4160116 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700000383.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK049334.1,BC018186.1,NM_028009.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rpusd1

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

pseudouridine synthesis
RNA modification
RNA binding
pseudouridine synthase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700003190.mm.1 NM_028009.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700003190.mm.1 NM_028009.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700003190.mm.1 TC1700000383.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700003189.mm.1 NM_028009.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700003189.mm.1 NM_028009.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700003189.mm.1 TC1700000383.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700003188.mm.1 NM_028009.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700003188.mm.1 NM_028009.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700003188.mm.1 TC1700000383.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 732238 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0