This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00551254_m1
See other PHYKPL GE Assays ›
Gene Symbol:
Phykpl
Gene Name:
5-phosphohydroxy-L-lysine phospholyase
Gene Aliases:
2900006B13Rik, Agxt2l2
Chromosome Location:
Chr.11: 51583983 - 51603269 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00551254_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Phykpl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028398.2 NP_082674.1 1 - 2 126 88
GenBank mRNA AK013489.1 - 1 - 2 80 88
AK085984.1 - 1 - 2 77 88
AK133235.1 - 73 88
AK156866.1 - 1 - 2 127 88
AK157759.1 - 1 - 2 76 88
AK160557.1 - 1 - 2 95 88
BC024461.1 - 1 - 2 80 4975449 88
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK087703.1 - 73 Chr. - - [-] on Build 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100000571.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Phykpl

Gene Ontology Categories:

Function(s) Process(es)

biological_process
catalytic activity
transaminase activity
transferase activity
lyase activity
pyridoxal phosphate binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100004430.mm.1 NM_028398.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100004430.mm.1 NM_028398.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100004430.mm.1 TC1100000571.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100004433.mm.1 NM_028398.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100004433.mm.1 NM_028398.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100004433.mm.1 TC1100000571.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100004431.mm.1 NM_028398.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100004431.mm.1 NM_028398.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100004431.mm.1 TC1100000571.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100004426.mm.1 NM_028398.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100004426.mm.1 NM_028398.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100004426.mm.1 TC1100000571.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 551369 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0