This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00551355_m1
See other BATF2 GE Assays ›
Gene Symbol:
Batf2
Gene Name:
basic leucine zipper transcription factor, ATF-like 2
Gene Aliases:
4933430F08Rik
Chromosome Location:
Chr.19: 6164392 - 6172476 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00551355_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Batf2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028967.1 NP_083243.1 1 - 2 98 87
GenBank mRNA AK150133.1 - 1 - 2 118 87
AK171828.1 - 1 - 2 118 87
BC024521.1 - 1 - 2 98 4984403 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000102.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK150133.1,AK171828.1,BC024521.1,NM_028967.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Batf2

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cell differentiation
defense response to protozoan
myeloid dendritic cell differentiation
DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900001187.mm.1 NM_028967.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900001187.mm.1 NM_028967.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900001187.mm.1 TC1900000102.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900001182.mm.1 NM_028967.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900001182.mm.1 NM_028967.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900001182.mm.1 TC1900000102.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900001181.mm.1 NM_028967.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900001181.mm.1 NM_028967.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900001181.mm.1 TC1900000102.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline