| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300016382.mm.1 |
NM_001170669.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016382.mm.1 |
NM_001170669.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016382.mm.1 |
NM_172263.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016382.mm.1 |
NM_172263.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016382.mm.1 |
TC1300002453.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300016383.mm.1 |
NM_001170669.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016383.mm.1 |
NM_001170669.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016383.mm.1 |
NM_172263.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016383.mm.1 |
NM_172263.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016383.mm.1 |
TC1300002453.mm.1 |
MTA TC |