This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00552109_m1
See other PDE8B GE Assays ›
Gene Symbol:
Pde8b
Gene Name:
phosphodiesterase 8B
Gene Aliases:
B230331L10Rik, C030047E14Rik
Chromosome Location:
Chr.13: 95024105 - 95250397 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00552109_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pde8b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001170669.1 NP_001164140.1 10 - 11 1305 72
NM_172263.2 NP_758467.2 11 - 12 1038 72
GenBank mRNA AK036090.1 - 13 - 14 1375 72
AK043680.1 - 10 - 11 1298 72
AK045991.1 - 10 - 11 1307 72
AK083166.1 - 10 - 11 943 72
BC032286.1 - 11 - 12 1050 5367055 72

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pde8b

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

phosphorelay signal transduction system
behavioral fear response
signal transduction
visual learning
operant conditioning
negative regulation of insulin secretion
neuromuscular process controlling balance
negative regulation of steroid hormone biosynthetic process
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
signal transducer activity
phosphoric diester hydrolase activity
hydrolase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline