This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00553280_g1
See other POLRMT GE Assays ›
Gene Symbol:
Polrmt
Gene Name:
polymerase (RNA) mitochondrial (DNA directed)
Gene Aliases:
1110018N15Rik, 4932416K13
Chromosome Location:
Chr.10: 79736125 - 79746589 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00553280_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polrmt
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172551.3 NP_766139.2 10 - 11 2587 64
GenBank mRNA AK003792.2 - 1 - 2 325 64
AK030039.1 - 9 - 10 2369 64
AK042071.1 - 8 - 9 2144 64
AK047545.1 - 10 - 11 2580 64
AK053356.1 - 10 - 11 2578 64
AK085666.1 - 10 - 11 2580 64
AK128956.1 - 11 - 12 2403 64
AK154181.1 - 10 - 11 2578 64
AK154729.1 - 10 - 11 2569 64
BC110697.1 - 10 - 11 2563 30356222 64
BC117939.1 - 10 - 11 2580 64
BC117940.1 - 10 - 11 2580 40097816 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002477.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Polrmt

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
transcription from mitochondrial promoter
DNA binding
DNA-directed RNA polymerase activity
transferase activity
nucleotidyltransferase activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000018099.mm.1 NM_172551.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000018099.mm.1 NM_172551.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000018099.mm.1 TC1000002477.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000018100.mm.1 NM_172551.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000018100.mm.1 NM_172551.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000018100.mm.1 TC1000002477.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0