This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00555351_g1
See other ATRIP GE Assays ›
Gene Symbol:
Atrip
Gene Name:
ATR interacting protein
Gene Aliases:
6620401K05Rik
Chromosome Location:
Chr.9: 109059747 - 109075580 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00555351_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atrip
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172774.3 NP_766362.2 7 - 8 1178 65
GenBank mRNA AK031257.1 - 7 - 8 1168 65
AK171423.1 - 7 - 8 1133 65
BC078458.1 - 7 - 8 1125 6832436 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900003073.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atrip

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
cellular response to DNA damage stimulus
protein binding
K63-linked polyubiquitin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900025901.mm.1 NM_172774.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900025901.mm.1 NM_172774.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900025901.mm.1 TC0900003073.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0