This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00555705_m1
See other PPWD1 GE Assays ›
Gene Symbol:
Ppwd1
Gene Name:
peptidylprolyl isomerase domain and WD repeat containing 1
Gene Aliases:
4632422M10Rik, A330090G21Rik
Chromosome Location:
Chr.13: 104204869 - 104228844 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00555705_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppwd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172807.4 NP_766395.2 5 - 6 1017 80
GenBank mRNA AK028532.1 - 5 - 6 997 80
BC037196.1 - 1 - 2 152 80
BC119282.1 - 5 - 6 995 8734031 80
BC119310.1 - 5 - 6 995 8734059 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002597.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppwd1

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
mRNA processing
protein folding
biological_process
RNA splicing
molecular_function
peptidyl-prolyl cis-trans isomerase activity
isomerase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300017699.mm.1 NM_172807.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017699.mm.1 NM_172807.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300017699.mm.1 TC1300002597.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0