This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00555874_m1
See other ADGRG7 GE Assays ›
Gene Symbol:
Adgrg7
Gene Name:
adhesion G protein-coupled receptor G7
Gene Aliases:
9130020O16Rik, Gpr128
Chromosome Location:
Chr.16: 56724609 - 56796021 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00555874_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adgrg7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172825.3 NP_766413.2 2 - 3 422 79
GenBank mRNA AK033593.1 - 2 - 3 423 79
AY255604.1 - 2 - 3 204 79
BC132302.1 - 2 - 3 247 40130579 79
BC137963.1 - 2 - 3 247 8860982 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001751.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adgrg7

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
biological_process
molecular_function
signal transducer activity
transmembrane signaling receptor activity
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600014227.mm.1 NM_172825.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600014227.mm.1 NM_172825.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600014227.mm.1 TC1600001751.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600014224.mm.1 NM_172825.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600014224.mm.1 NM_172825.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600014224.mm.1 TC1600001751.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600014225.mm.1 NM_172825.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600014225.mm.1 NM_172825.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600014225.mm.1 TC1600001751.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600014226.mm.1 NM_172825.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600014226.mm.1 NM_172825.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600014226.mm.1 TC1600001751.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0