This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00556137_m1
See other CDH7 GE Assays ›
Gene Symbol:
Cdh7
Gene Name:
cadherin 7, type 2
Gene Aliases:
9330156F07Rik, CDH7L1
Chromosome Location:
Chr.1: 109982327 - 110141917 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00556137_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdh7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172853.2 NP_766441.1 4 - 5 974 64
GenBank mRNA AK034096.1 - 4 - 5 974 64
AK042657.1 - 6 - 7 1369 64
AK137369.1 - 4 - 5 897 64
AY267034.1 - 3 - 4 829 64
BC083189.1 - 5 - 6 1564 64
BC141119.1 - 3 - 4 805 9055731 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100001037.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdh7

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
calcium ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100008633.mm.1 NM_172853.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100008633.mm.1 TC0100001037.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100008631.mm.1 NM_172853.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100008631.mm.1 TC0100001037.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100008634.mm.1 NM_172853.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100008634.mm.1 TC0100001037.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline