This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00556225_g1
See other FREM2 GE Assays ›
Gene Symbol:
Frem2
Gene Name:
Fras1 related extracellular matrix protein 2
Gene Aliases:
6030440P17Rik, 8430406N05Rik, Gm409, b2b1562Clo, my, ne, nv1
Chromosome Location:
Chr.3: 53513938 - 53657912 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00556225_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Frem2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172862.3 NP_766450.2 19 - 20 8649 70
GenBank mRNA AB236662.1 - 19 - 20 8649 70
AJ833643.1 - 19 - 20 8378 70
AK031494.1 - 9 - 10 1551 70
AK082256.1 - 2 - 3 679 70
BC068185.1 - 9 - 10 1551 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300001983.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK082256.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Frem2

Gene Ontology Categories:

Function(s) Process(es)

morphogenesis of an epithelium
cell communication
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
multicellular organism development
inner ear development
calcium ion binding
protein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300015062.mm.1 NM_172862.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015062.mm.1 NM_172862.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300015062.mm.1 TC0300001983.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300015061.mm.1 NM_172862.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015061.mm.1 NM_172862.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300015061.mm.1 TC0300001983.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-48sk5:80/100.66.77.18:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0