This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00557070_m1
See other PHTF2 GE Assays ›
Gene Symbol:
Phtf2
Gene Name:
putative homeodomain transcription factor 2
Gene Aliases:
1110054G21Rik, 9530062N20Rik, AI447096, C77923
Chromosome Location:
Chr.5: 20758664 - 20882124 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00557070_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Phtf2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172992.3 NP_766580.3 13 - 14 1960 114
GenBank mRNA AK037021.1 - 13 - 14 1847 114
AK162226.1 - 2 - 3 242 114
AK163376.1 - 13 - 14 1887 114
BC052841.1 - 13 - 14 1873 30050876 114

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002007.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Phtf2

Panther Classification:

Molecular Function -

homeodomain transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
molecular_function
DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500018218.mm.1 TC0500002007.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500018218.mm.1 NM_172992.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500018218.mm.1 NM_172992.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0