This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00557149_g1
See other MON1B GE Assays ›
Gene Symbol:
Mon1b
Gene Name:
MON1 homolog b (yeast)
Gene Aliases:
5031407H10, 5033413H12Rik
Chromosome Location:
Chr.8: 113635586 - 113645192 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00557149_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mon1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001048143.2 NP_001041608.1 3 - 4 622 117
NM_173015.3 NP_766603.1 2 - 3 812 117
GenBank mRNA AK030288.1 - 2 - 3 778 117
AK157390.1 - 3 - 4 622 117
AK173047.1 - 2 - 3 484 117
BC062639.1 - 2 - 3 488 117
CA320533.1 - 2 - 3 402 117
CJ089261.1 - 3 - 4 623 117

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001357.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mon1b

Gene Ontology Categories:

Function(s) Process(es)

biological_process
vesicle-mediated transport
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800011134.mm.1 NM_001048143.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800011134.mm.1 NM_173015.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800011134.mm.1 TC0800001357.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0