This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00558179_m1
See other ATG4C GE Assays ›
Gene Symbol:
Atg4c
Gene Name:
autophagy related 4C, cysteine peptidase
Gene Aliases:
Apg4-C, Apg4c, Atg4cl, Autl1
Chromosome Location:
Chr.4: 99190891 - 99297258 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00558179_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atg4c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001145967.1 NP_001139439.1 8 - 9 1209 80
NM_175029.3 NP_778194.3 8 - 9 1591 80
GenBank mRNA AK134838.1 - 8 - 9 1358 80
BC058981.1 - 8 - 9 1591 6827140 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400000996.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atg4c

Panther Classification:

Molecular Function -

cysteine protease

Gene Ontology Categories:

Function(s) Process(es)

autophagosome assembly
mitophagy
C-terminal protein lipidation
proteolysis
protein targeting to membrane
transport
autophagy
cellular response to nitrogen starvation
protein transport
protein processing
nucleophagy
protein delipidation
cysteine-type endopeptidase activity
peptidase activity
cysteine-type peptidase activity
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400006729.mm.1 NM_001145967.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400006729.mm.1 NM_001145967.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400006729.mm.1 NM_175029.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400006729.mm.1 NM_175029.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400006729.mm.1 TC0400000996.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0