This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00558667_g1
See other OSCAR GE Assays ›
Gene Symbol:
Oscar
Gene Name:
osteoclast associated receptor
Gene Aliases:
mOSCAR, mOSCAR-M1, mOSCAR-M2, mOSCAR-M3
Chromosome Location:
Chr.7: 3609813 - 3616157 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00558667_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Oscar
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290377.1 NP_001277306.1 4 - 5 730 107
NM_175632.3 NP_783440.1 4 - 5 748 107
GenBank mRNA AF391159.1 - 4 - 5 706 107
AF391160.1 - 4 - 5 730 107
AF391161.1 - 4 - 5 724 107
BC137776.1 - 4 - 5 675 8860795 107

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002131.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Oscar

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946359 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977426 AF391161.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977426 NM_175632.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977426 AF391159.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 641 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 674874 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 45952 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 41075 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 19452 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117448 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136593 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977426 AF391161.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977426 NM_175632.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977426 AF391159.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946334 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946355 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946337 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946344 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946339 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946340 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946332 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946343 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946336 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946351 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946347 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946350 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946357 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946358 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946353 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136593 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P117448 - -
Whole Mouse Genome G4122A A_51_P404478 BC137776.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P404478 AF391161.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P404478 NM_175632.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P404478 AF391159.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108014 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P117448 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136593 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946359 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946283 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946374 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946399 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946315 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946317 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946377 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946407 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946320 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946337 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946306 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946361 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946343 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946336 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946406 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946329 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946369 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946370 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946277 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946313 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946351 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946413 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946282 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946305 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946279 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946327 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946357 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946302 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946365 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946353 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946348 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946396 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946330 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946291 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946338 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946360 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946284 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946401 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946363 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946328 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946288 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946410 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946324 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946404 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946297 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946409 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946301 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946307 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946304 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946294 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946325 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946372 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946332 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946298 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946350 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946308 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946336 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946334 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946321 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946353 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946320 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946344 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946322 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946339 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946350 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946351 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946314 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946325 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946341 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946317 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946348 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946340 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946327 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946330 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946355 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946318 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946323 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946329 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946324 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946338 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946315 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946328 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946332 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946343 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24946337 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10559400 - -
Affymetrix Mouse Gene 1.0 ST Array 10559404 - -
Affymetrix Mouse Gene 1.0 ST Array 10559401 - -
Affymetrix Mouse Gene 1.0 ST Array 10559405 - -
Affymetrix Mouse Gene 1.0 ST Array 10559403 - -
Affymetrix Mouse Gene 1.0 ST Array 10559402 - -
Affymetrix HT MG-430 PM Array Plate 1451710_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1425442_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10559399 - -
Mouse Genome 430 2.0 Array 1425442_PM_at - -
Mouse Genome 430 2.0 Array 1425442_at - -
Mouse Genome 430 2.0 Array 1451710_PM_at - -
Mouse Genome 430 2.0 Array 1451710_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 232790_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 232790_PM_at - -
Affymetrix Mouse Clariom S Array PSR0700019173.mm.1 TC0700002131.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700019173.mm.1 NM_175632.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700019172.mm.1 TC0700002131.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700019172.mm.1 NM_175632.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700019174.mm.1 TC0700002131.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700019174.mm.1 NM_175632.3 Refseq
Mouse Expression Set 430 1451710_at - -
Mouse Expression Set 430 1451710_PM_at - -
Mouse Expression Set 430 1425442_PM_at - -
Mouse Expression Set 430 1425442_at - -
Mouse Genome 430A 2.0 Array 1451710_at - -
Mouse Genome 430A 2.0 Array 1451710_PM_at - -
Mouse Genome 430A 2.0 Array 1425442_at - -
Mouse Genome 430A 2.0 Array 1425442_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1425442_at - -
Affymetrix HT Mouse Genome 430A Array 1451710_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1425442_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1451710_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 2120441 - -
Illumina mouseRef-8 v1.0 expression beadchip 0002120441 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1215466 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1215466 - -
mouse-6 v1.1 (Illumina) scl31835.5.1_93-S - -
Illumina Sentrix mouse-6.v1 bead array 2120441 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl31835.5.1_93-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 2120441 - -
MouseRef-8 v2 Expression BeadChip ILMN_1215466 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215466 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0