This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00565144_m1
See other OPRM1 GE Assays ›
Gene Symbol:
Oprm1
Gene Name:
opioid receptor, mu 1
Gene Aliases:
M-OR-1, MOP-R, MOR-1, MOR-1O, Oprm, mor, muOR
Chromosome Location:
Chr.10: 6758593 - 7038209 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00565144_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Oprm1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AB047546.1 - 3 - 4 1379 68
AB441735.1 - 3 - 4 1165 68
AB441736.1 - 3 - 4 1165 68
AB441737.1 - 3 - 4 1165 68
AB441738.1 - 3 - 4 1165 68
AB441739.1 - 3 - 4 1165 68
AB441740.1 - 3 - 4 1165 68
AB441741.1 - 3 - 4 1165 68
AB441742.1 - 3 - 4 1165 68
AB441743.1 - 3 - 4 1165 68
AF062755.1 - 3 - 4 1066 68
AF260306.1 - 4 - 5 1394 68
AF260307.1 - 4 - 5 1568 68
AF260308.1 - 5 - 6 1523 68
AF260309.1 - 4 - 5 1128 68
AF260310.1 - 4 - 5 1087 68
AF400246.1 - 4 - 5 1394 68
AF400247.1 - 4 - 5 1568 68
AF400248.1 - 5 - 6 1523 68
AY358080.1 - 1 - 2 528 68
AY923184.2 - 5 - 6 1497 68
EF105313.1 - 3 - 4 1231 68
HQ699464.1 - 2 - 3 878 68
U19380.1 - 3 - 4 1420 68
U26915.1 - 3 - 4 1447 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0