This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00600340_m1
See other ECHDC3 GE Assays ›
Gene Symbol:
Echdc3
Gene Name:
enoyl Coenzyme A hydratase domain containing 3
Gene Aliases:
2310005D12Rik, AI662097
Chromosome Location:
Chr.2: 6188465 - 6212994 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00600340_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Echdc3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024208.4 NP_077170.2 3 - 4 424 76
GenBank mRNA AK004218.1 - 3 - 4 426 76
AK009166.1 - 3 - 4 425 76
BC002214.1 - 3 - 4 405 3490443 76
BC054365.1 - 3 - 4 393 5343451 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002890.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Echdc3

Gene Ontology Categories:

Function(s) Process(es)

biological_process
metabolic process
molecular_function
catalytic activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200024021.mm.1 NM_024208.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200024021.mm.1 NM_024208.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200024021.mm.1 TC0200002890.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0