This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00600351_m1
See other HAGH GE Assays ›
Gene Symbol:
Hagh
Gene Name:
hydroxyacyl glutathione hydrolase
Gene Aliases:
BC019817, Glo-2, Glo2, Rsp29
Chromosome Location:
Chr.17: 24840079 - 24864450 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00600351_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hagh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159626.1 NP_001153098.1 7 - 8 765 98
NM_024284.2 NP_077246.2 7 - 8 855 98
GenBank mRNA AK147022.1 - 7 - 8 816 98
AK150557.1 - 7 - 8 773 98
AK151231.1 - 7 - 8 773 98
AK152742.1 - 7 - 8 773 98
AK165777.1 - 7 - 8 759 98
BC004749.1 - 7 - 8 798 3583919 98
BC019817.1 - 7 - 8 748 5123648 98
BQ128470.1 - 2 - 3 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700000354.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hagh

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
glutathione metabolic process
glutathione biosynthetic process
hydroxyacylglutathione hydrolase activity
zinc ion binding
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700002795.mm.1 NM_001159626.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700002795.mm.1 NM_001159626.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700002795.mm.1 NM_024284.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700002795.mm.1 NM_024284.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700002795.mm.1 TC1700000354.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0