This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00613556_m1
See other SLC25A16 GE Assays ›
Gene Symbol:
Slc25a16
Gene Name:
solute carrier family 25 (mitochondrial carrier, Graves disease autoantigen), member 16
Gene Aliases:
3110021G18Rik, GDA, GDC, HGT.1, ML7
Chromosome Location:
Chr.10: 62920633 - 62946499 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00613556_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc25a16
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175194.2 NP_780403.1 2 - 3 347 83
GenBank mRNA AK030865.1 - 2 - 3 347 83
BC025015.1 - 2 - 3 258 83
BC138983.1 - 2 - 3 269 8862005 83
BC138984.1 - 2 - 3 269 8862006 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000590.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc25a16

Panther Classification:

Molecular Function -

mitochondrial carrier protein

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
transmembrane transport
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000003967.mm.1 NM_175194.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003967.mm.1 NM_175194.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000003967.mm.1 TC1000000590.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0