This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00614084_g1
See other BORA GE Assays ›
Gene Symbol:
Bora
Gene Name:
bora, aurora kinase A activator
Gene Aliases:
6720463M24Rik, AI317232
Chromosome Location:
Chr.14: 99046247 - 99076450 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00614084_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bora
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175265.4 NP_780474.1 3 - 4 571 112
GenBank mRNA AK034930.1 - 3 - 4 572 112
AK141389.1 - 3 - 4 522 112
AK167430.1 - 3 - 4 503 112
BC052183.1 - 3 - 4 472 6336038 112

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bora

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
mitotic nuclear division
regulation of mitotic nuclear division
regulation of protein localization
cell division
regulation of mitotic spindle organization
protein kinase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400009057.mm.1 NM_175265.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009057.mm.1 NM_175265.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400009057.mm.1 TC1400001152.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400009058.mm.1 NM_175265.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009058.mm.1 NM_175265.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400009058.mm.1 TC1400001152.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0