This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00614447_m1
See other SLCO2B1 GE Assays ›
Gene Symbol:
Slco2b1
Gene Name:
solute carrier organic anion transporter family, member 2b1
Gene Aliases:
AI060904, AI852653, OATP-B, Slc21a9
Chromosome Location:
Chr.7: 99657804 - 99711340 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00614447_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slco2b1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252530.1 NP_001239459.1 8 - 9 1382 95
NM_001252531.1 NP_001239460.1 8 - 9 1325 95
NM_175316.3 NP_780525.1 8 - 9 1128 95
GenBank mRNA AK041144.1 - 8 - 9 1324 95
AK048120.1 - 8 - 9 1129 95
AK087682.1 - 8 - 9 1380 95
AK132394.1 - 8 - 9 1383 95
AK163542.1 - 8 - 9 1378 95
AK173049.1 - 6 - 7 914 95
BC019209.1 - 8 - 9 1111 95
BC037089.1 - 8 - 9 1100 95
BC049251.1 - 8 - 9 1091 95
BC096485.1 - 8 - 9 1137 5101188 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700003804.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slco2b1

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
organic anion transport
bile acid and bile salt transport
sodium-independent icosanoid transport
anion transmembrane transport
organic acid transmembrane transport
transporter activity
organic anion transmembrane transporter activity
bile acid transmembrane transporter activity
sodium-independent organic anion transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700032017.mm.1 NM_001252530.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700032017.mm.1 NM_001252530.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700032017.mm.1 NM_001252531.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700032017.mm.1 NM_001252531.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700032017.mm.1 NM_175316.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700032017.mm.1 NM_175316.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700032017.mm.1 TC0700003804.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline