This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00614796_m1
See other UNC119B GE Assays ›
Gene Symbol:
Unc119b
Gene Name:
unc-119 homolog B (C. elegans)
Gene Aliases:
AA407659, AI414892
Chromosome Location:
Chr.5: 115122566 - 115134975 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00614796_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Unc119b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175352.4 NP_780561.1 2 - 3 420 97
GenBank mRNA AK082614.1 - 2 - 3 422 97
BC112388.1 - 2 - 3 392 5352914 97
BC115529.2 - 2 - 3 374 40101389 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003089.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Unc119b

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

transport
protein transport
cell projection organization
lipoprotein transport
cilium morphogenesis
lipid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500027469.mm.1 TC0500003089.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500027469.mm.1 NM_175352.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500027469.mm.1 NM_175352.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jmd5k:80/100.66.78.41:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0