This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00615264_m1
See other ALDH4A1 GE Assays ›
Gene Symbol:
Aldh4a1
Gene Name:
aldehyde dehydrogenase 4 family, member A1
Gene Aliases:
A930035F14Rik, Ahd-1, Ahd1, Aldh4, Aldh5a1, E330022C09, P5cd, P5cdh, P5cdhl, P5cdhs, Ssdh1
Chromosome Location:
Chr.4: 139622871 - 139649691 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00615264_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aldh4a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175438.4 NP_780647.3 3 - 4 488 61
GenBank mRNA AK044712.1 - 3 - 4 284 61
BC024133.1 - 2 - 3 206 61
BC039281.1 - 3 - 4 272 4975491 61
BC056226.1 - 3 - 4 372 5064715 61
BC060650.1 - 3 - 4 488 30354228 61
CA494657.1 - 3 - 4 525 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400004196.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Aldh4a1

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

glutamate biosynthetic process
proline metabolic process
proline biosynthetic process
metabolic process
oxidation-reduction process
1-pyrroline-5-carboxylate dehydrogenase activity
aldehyde dehydrogenase (NAD) activity
oxidoreductase activity
oxidoreductase activity, acting on the aldehyde or oxo group of donors, NAD or NADP as acceptor
identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400014478.mm.1 NM_175438.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400014478.mm.1 NM_175438.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400014478.mm.1 TC0400004196.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0