This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00617659_g1
See other PRSS33 GE Assays ›
Gene Symbol:
Prss33
Gene Name:
protease, serine 33
Gene Aliases:
Eos, mT6, tryptase-6
Chromosome Location:
Chr.17: 23833338 - 23836126 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00617659_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prss33
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081399.2 NP_001074868.2 3 - 4 276 84
GenBank mRNA AY262280.1 - 3 - 4 252 84
BC101950.1 - 3 - 4 252 40044230 84
BC103541.1 - 4 - 5 348 40044224 84
BC103542.1 - 3 - 4 252 40044219 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001607.mm.1

Set Membership:

Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700014087.mm.1 TC1700001607.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700014082.mm.1 NM_001081399.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700014082.mm.1 NM_001081399.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700014082.mm.1 TC1700001607.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-lsr54:80/100.66.79.61:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0