This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00618108_m1
See other LAIR1 GE Assays ›
Gene Symbol:
Lair1
Gene Name:
leukocyte-associated Ig-like receptor 1
Gene Aliases:
5133400O11Rik, BB115266, D7Bwg0421e, Lair-1
Chromosome Location:
Chr.7: 4007073 - 4073507 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00618108_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lair1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178611.6 NP_848726.1 1 - 2 135 89
GenBank mRNA AF491830.1 - 1 - 2 47 89
AK017222.1 - 1 - 2 139 89
AK033472.1 - 1 - 2 112 89
AK038574.1 - 1 - 2 98 89
AY392764.1 - 1 - 2 72 89

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF491830.1, AK017222.1, AK033472.1, AK038574.1, AY392764.1, NM_178611.6 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lair1

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0