This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00618112_g1
See other LAIR1 GE Assays ›
Gene Symbol:
Lair1
Gene Name:
leukocyte-associated Ig-like receptor 1
Gene Aliases:
5133400O11Rik, BB115266, D7Bwg0421e, Lair-1
Chromosome Location:
Chr.7: 4007073 - 4073507 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00618112_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lair1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113474.1 NP_001106945.1 6 - 7 701 94
NM_178611.6 NP_848726.1 5 - 6 371 94
GenBank mRNA AF479685.1 - 6 - 7 636 94
AF491829.1 - 6 - 7 734 94
AF491830.1 - 5 - 6 283 94
AF491831.1 - 6 - 7 613 94
AK017222.1 - 5 - 6 375 94
AK033472.1 - 5 - 6 348 94
AK038574.1 - 5 - 6 334 94
AK043664.1 - 5 - 6 428 94
AK044465.1 - 6 - 7 672 94
AK050281.1 - 6 - 7 657 94
AY392763.1 - 6 - 7 638 94
AY392764.1 - 5 - 6 308 94
AY392765.1 - 4 - 5 191 94
AY392766.1 - 5 - 6 521 94
BC025901.1 - 6 - 7 665 5253846 94
BY204263.1 - 4 - 5 315 94

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lair1

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0