Assay Details
| Gene Symbol: Slc30a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178651.3 | NP_848766.2 | 10 - 11 | 1036 | 109 | |
| GenBank mRNA | AF263460.1 | - | 8 - 9 | 590 | 109 | |
| AK028885.1 | - | 10 - 11 | 1003 | 109 | ||
| AK033990.1 | - | 61 | 109 | |||
| AK035541.1 | - | 10 - 11 | 1023 | 109 | ||
| AK038559.1 | - | 9 - 10 | 942 | 109 | ||
| AK082853.1 | - | 10 - 11 | 1038 | 109 | ||
| AK143277.1 | - | 10 - 11 | 1037 | 109 | ||
| AK149549.1 | - | 3 - 4 | 1187 | 109 | ||
| AK179008.1 | - | 2 - 3 | 109 | |||
| AK182342.1 | - | 2 - 3 | 114 | 109 | ||
| AK205884.1 | - | 2 - 3 | 82 | 109 | ||
| AY682914.1 | - | 11 - 12 | 1029 | 109 | ||
| BC023833.1 | - | 2 - 3 | 140 | 109 | ||
| BC026565.1 | - | 6 - 7 | 454 | 109 | ||
| BC027806.1 | - | 10 - 11 | 798 | 109 | ||
| BC031705.1 | - | 7 - 8 | 550 | 109 | ||
| BC055773.1 | - | 7 - 8 | 489 | 109 | ||
| BC078440.1 | - | 8 - 9 | 613 | 109 | ||
| BC141008.1 | - | 11 - 12 | 1002 | 9055620 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc30a9
Genomic Map