This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00618359_m1
See other RBMS3 GE Assays ›
Gene Symbol:
Rbms3
Gene Name:
RNA binding motif, single stranded interacting protein
Gene Aliases:
6720477E09Rik, 8430436O14Rik
Chromosome Location:
Chr.9: 116572746 - 118040292 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00618359_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbms3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001172121.1 NP_001165592.1 9 - 10 983 71
NM_001172122.1 NP_001165593.1 8 - 9 1455 71
NM_001172123.1 NP_001165594.1 8 - 9 1455 71
NM_001172124.1 NP_001165595.1 8 - 9 1455 71
NM_001172126.1 NP_001165597.1 8 - 9 1455 71
NM_178660.4 NP_848775.2 8 - 9 1455 71
GenBank mRNA AK030478.1 - 8 - 9 1115 71
AK030609.1 - 8 - 9 1089 71
AK032938.1 - 8 - 9 1452 71
AK033005.1 - 8 - 9 1098 71
AK041098.1 - 8 - 9 1457 71
AK046197.1 - 9 - 10 984 71
AK051776.1 - 9 - 10 1502 71
AK052196.1 - 8 - 9 1436 71
BC033604.1 - 4 - 5 310 71
BC108402.1 - 8 - 9 1429 6516420 71
BC117841.1 - 8 - 9 895 40093225 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900003204.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rbms3

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of translation
nucleotide binding
nucleic acid binding
RNA binding
mRNA 3'-UTR binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900027025.mm.1 NM_001172121.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027025.mm.1 NM_001172121.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027025.mm.1 NM_001172122.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027025.mm.1 NM_001172122.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027025.mm.1 NM_001172123.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027025.mm.1 NM_001172123.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027025.mm.1 NM_001172124.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027025.mm.1 NM_001172124.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027025.mm.1 NM_001172126.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027025.mm.1 NM_001172126.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027025.mm.1 NM_178660.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027025.mm.1 NM_178660.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900027025.mm.1 TC0900003204.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-bjcbn:80/100.66.75.54:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0