This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00618361_m1
See other RBMS3 GE Assays ›
Gene Symbol:
Rbms3
Gene Name:
RNA binding motif, single stranded interacting protein
Gene Aliases:
6720477E09Rik, 8430436O14Rik
Chromosome Location:
Chr.9: 116572746 - 118040292 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00618361_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbms3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001172121.1 NP_001165592.1 11 - 12 1189 117
NM_001172123.1 NP_001165594.1 11 - 12 1712 117
NM_001172124.1 NP_001165595.1 11 - 12 1712 117
NM_178660.4 NP_848775.2 10 - 11 1661 117
GenBank mRNA AK030478.1 - 10 - 11 1321 117
AK032938.1 - 10 - 11 1658 117
AK033005.1 - 11 - 12 1355 117
AK046197.1 - 11 - 12 1190 117
AK052196.1 - 11 - 12 1693 117
BC108402.1 - 10 - 11 1635 6516420 117
BC117841.1 - 10 - 11 1101 40093225 117

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900003204.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rbms3

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of translation
nucleotide binding
nucleic acid binding
RNA binding
mRNA 3'-UTR binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900027020.mm.1 NM_001172121.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027020.mm.1 NM_001172121.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027020.mm.1 NM_001172123.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027020.mm.1 NM_001172123.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027020.mm.1 NM_001172124.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027020.mm.1 NM_001172124.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027020.mm.1 NM_178660.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027020.mm.1 NM_178660.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900027020.mm.1 TC0900003204.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-lsr54:80/100.66.79.61:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0