This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00618535_g1
See other RAB3GAP1 GE Assays ›
Gene Symbol:
Rab3gap1
Gene Name:
RAB3 GTPase activating protein subunit 1
Gene Aliases:
1700003B17Rik, 4732493F09Rik, AL117896, Rab3gap, p130
Chromosome Location:
Chr.1: 127868737 - 127943876 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00618535_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rab3gap1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178690.4 NP_848805.2 19 - 20 2333 83
GenBank mRNA AK076244.1 - 19 - 20 2297 83
AK076399.1 - 19 - 20 2316 83
AK080432.1 - 19 - 20 2315 83
AK134604.1 - 19 - 20 2315 83
AK164523.1 - 19 - 20 2314 83
AK170768.1 - 19 - 20 2306 83
AK172881.1 - 19 - 20 2320 83
BC028996.1 - 3 - 4 741 83
BC038485.1 - 3 - 4 647 83
BC046297.1 - 19 - 20 2299 6486170 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100001125.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rab3gap1

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

brain development
positive regulation of gene expression
regulation of synaptic vesicle priming
hypothalamus development
lipid particle organization
camera-type eye development
regulation of GTPase activity
positive regulation of GTPase activity
regulation of short-term neuronal synaptic plasticity
synaptic vesicle transport
protein stabilization
excitatory postsynaptic potential
face morphogenesis
positive regulation of glutamate neurotransmitter secretion in response to membrane depolarization
establishment of protein localization to endoplasmic reticulum membrane
regulation of calcium ion-dependent exocytosis of neurotransmitter
positive regulation of endoplasmic reticulum tubular network organization
GTPase activator activity
Rab guanyl-nucleotide exchange factor activity
Rab GTPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100009137.mm.1 NM_178690.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100009137.mm.1 NM_178690.4 Refseq
Affymetrix Mouse Clariom S Array PSR0100009137.mm.1 TC0100001125.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0