This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00618777_m1
See other CADM2 GE Assays ›
Gene Symbol:
Cadm2
Gene Name:
cell adhesion molecule 2
Gene Aliases:
2900078E11Rik, 9330131D06, A830029E02Rik, Igdf4d, Igsf4d, NECL3, SynCAM 2, SynCAM2
Chromosome Location:
Chr.16: 66655421 - 67621200 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00618777_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cadm2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001145977.1 NP_001139449.1 3 - 4 1037 76
NM_178721.4 NP_848836.2 4 - 5 1064 76
GenBank mRNA AK033973.1 - 3 - 4 664 76
AK038842.1 - 4 - 5 1043 76
AK043760.1 - 4 - 5 1047 76
AK046800.1 - 3 - 4 940 76
AK163636.1 - 4 - 5 1041 76
AK165030.1 - 4 - 5 1061 76
BC139401.1 - 4 - 5 633 8862423 76
BC139402.1 - 4 - 5 633 8862424 76

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cadm2

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0