This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00619440_m1
See other GTF2H3 GE Assays ›
Gene Symbol:
Gtf2h3
Gene Name:
general transcription factor IIH, polypeptide 3
Gene Aliases:
34kDa, 5033417D07Rik, BTF2, BTF2 p34, C730029A10, D5Ertd679e, TFIIH
Chromosome Location:
Chr.5: 124579148 - 124597680 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00619440_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gtf2h3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_181410.3 NP_852075.1 5 - 6 467 121
GenBank mRNA AK050227.1 - 5 - 6 451 121
AK133941.1 - 5 - 6 438 121
BC017515.1 - 5 - 6 444 4504077 121

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001449.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gtf2h3

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
nucleotide-excision repair
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
translation
cellular response to DNA damage stimulus
phosphorylation of RNA polymerase II C-terminal domain
damaged DNA binding
protein kinase activity
DNA-dependent ATPase activity
translation factor activity, RNA binding
RNA polymerase II carboxy-terminal domain kinase activity
metal ion binding
protein N-terminus binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500013022.mm.1 NM_181410.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500013022.mm.1 NM_181410.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500013022.mm.1 TC0500001449.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500013023.mm.1 NM_181410.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500013023.mm.1 NM_181410.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500013023.mm.1 TC0500001449.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline