This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00619526_g1
See other ARHGAP11A GE Assays ›
Gene Symbol:
Arhgap11a
Gene Name:
Rho GTPase activating protein 11A
Gene Aliases:
6530401L14Rik, mKIAA0013
Chromosome Location:
Chr.2: 113831492 - 113848661 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00619526_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap11a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_181416.3 NP_852081.2 5 - 6 1289 78
GenBank mRNA AK129034.1 - 1284 78
AK137111.1 - 5 - 6 926 78
BC050875.1 - 5 - 6 1243 6511064 78
BC058105.1 - 5 - 6 1289 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200004366.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arhgap11a

Panther Classification:

Molecular Function -

G-protein modulator protein-binding activity modulator GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
molecular_function
GTPase activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200036478.mm.1 NM_181416.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200036478.mm.1 NM_181416.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200036478.mm.1 TC0200004366.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200036476.mm.1 NM_181416.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200036476.mm.1 NM_181416.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200036476.mm.1 TC0200004366.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200036477.mm.1 NM_181416.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200036477.mm.1 NM_181416.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200036477.mm.1 TC0200004366.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0