This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00622070_m1
See other PTPN7 GE Assays ›
Gene Symbol:
Ptpn7
Gene Name:
protein tyrosine phosphatase, non-receptor type 7
Gene Aliases:
BPTP-4, C920001D21Rik, HEPTP, LC-PTP, LPTP
Chromosome Location:
Chr.1: 135131840 - 135145328 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00622070_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ptpn7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177081.3 NP_796055.1 1 - 2 70 81
GenBank mRNA AK083305.1 - 1 - 2 70 81

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK083305.1, NM_177081.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ptpn7

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
dephosphorylation
peptidyl-tyrosine dephosphorylation
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
hydrolase activity
phosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100010218.mm.1 NM_177081.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100010218.mm.1 NM_177081.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100010218.mm.1 TC0100001244.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100010220.mm.1 NM_177081.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100010220.mm.1 NM_177081.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100010220.mm.1 TC0100001244.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0