This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00624160_m1
See other SERAC1 GE Assays ›
Gene Symbol:
Serac1
Gene Name:
serine active site containing 1
Gene Aliases:
4930511N22Rik, D17Ertd141e
Chromosome Location:
Chr.17: 6040571 - 6079762 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00624160_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Serac1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001111017.1 NP_001104487.1 6 - 7 605 72
NM_177311.4 NP_796285.2 5 - 6 515 72
GenBank mRNA AK019677.1 - 5 - 6 504 72
AK154605.1 - 6 - 7 556 72
AK155571.1 - 6 - 7 608 72
AK162246.1 - 6 - 7 540 72
AK170498.1 - 6 - 7 556 72
BC119594.1 - 6 - 7 594 40059066 72
BC119595.1 - 6 - 7 594 40059071 72
BY715309.1 - 5 - 6 501 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001378.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Serac1

Gene Ontology Categories:

Function(s) Process(es)

GPI anchor metabolic process
lipid metabolic process
intracellular protein transport
phospholipid biosynthetic process
extracellular matrix organization
hydrolase activity, acting on ester bonds

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700012401.mm.1 NM_001111017.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700012401.mm.1 NM_001111017.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700012401.mm.1 NM_177311.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700012401.mm.1 NM_177311.4 Refseq
Affymetrix Mouse Clariom S Array PSR1700012401.mm.1 TC1700001378.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7xkh5:80/100.66.78.74:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0