This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00624388_m1
See other KAT7 GE Assays ›
Gene Symbol:
Kat7
Gene Name:
K(lysine) acetyltransferase 7
Gene Aliases:
Hbo1, Hboa, Myst2
Chromosome Location:
Chr.11: 95271853 - 95310246 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00624388_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Kat7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001195003.1 NP_001181932.1 4 - 5 772 98
NM_001195004.1 NP_001181933.1 4 - 5 772 98
NM_177619.3 NP_808287.1 3 - 4 595 98
GenBank mRNA AK147968.1 - 3 - 4 499 98
AK169458.1 - 4 - 5 694 98
AK170389.1 - 4 - 5 719 98
BC024092.1 - 2 - 3 345 98
BC048904.1 - 3 - 4 550 6492124 98
BC057102.1 - 4 - 5 700 6401353 98
DQ076247.1 - 4 - 5 700 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003588.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Kat7

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

DNA replication
transcription, DNA-templated
regulation of transcription, DNA-templated
chromatin modification
histone H3 acetylation
histone H4-K5 acetylation
histone H4-K8 acetylation
histone H4-K12 acetylation
transcription factor activity, sequence-specific DNA binding
histone acetyltransferase activity
zinc ion binding
transferase activity
transferase activity, transferring acyl groups
transferase activity, transferring acyl groups other than amino-acyl groups
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100032918.mm.1 NM_001195003.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032918.mm.1 NM_001195003.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032918.mm.1 NM_001195004.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032918.mm.1 NM_001195004.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032918.mm.1 NM_177619.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032918.mm.1 NM_177619.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100032918.mm.1 TC1100003588.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0