This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00625015_g1
See other AWAT2 GE Assays ›
Gene Symbol:
Awat2
Gene Name:
acyl-CoA wax alcohol acyltransferase 2
Gene Aliases:
9430062J17Rik, ARAT, Dgat2l4, WS
Chromosome Location:
Chr.X: 100399336 - 100494221 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00625015_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Awat2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290395.1 NP_001277324.1 5 - 6 891 119
NM_177746.4 NP_808414.2 4 - 5 521 119
GenBank mRNA AK034920.1 - 5 - 6 892 119
AY611031.1 - 4 - 5 496 119
AY611032.1 - 4 - 5 521 119
BC138085.1 - 4 - 5 489 8861104 119
BC138086.1 - 4 - 5 489 8861105 119

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00002703.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Awat2

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
wax biosynthetic process
molecular_function
transferase activity
transferase activity, transferring acyl groups
transferase activity, transferring acyl groups other than amino-acyl groups
long-chain-alcohol O-fatty-acyltransferase activity
retinol O-fatty-acyltransferase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00016950.mm.1 NM_177746.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00016950.mm.1 TC0X00002703.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 720400 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0