This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00625350_m1
See other VSIG4 GE Assays ›
Gene Symbol:
Vsig4
Gene Name:
V-set and immunoglobulin domain containing 4
Gene Aliases:
A530061A11, BC025105, CRIg, Z39IG
Chromosome Location:
Chr.X: 96247201 - 96293518 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00625350_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Vsig4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177789.4 NP_808457.1 5 - 6 713 83
GenBank mRNA AK041014.1 - 5 - 6 714 83
BC025105.1 - 5 - 6 689 4162875 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00002676.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Vsig4

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s)

negative regulation of interleukin-2 production
negative regulation of T cell proliferation

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00016786.mm.1 NM_177789.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00016786.mm.1 NM_177789.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00016786.mm.1 TC0X00002676.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00016782.mm.1 NM_177789.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00016782.mm.1 NM_177789.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00016782.mm.1 TC0X00002676.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00016783.mm.1 NM_177789.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00016783.mm.1 NM_177789.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00016783.mm.1 TC0X00002676.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00016784.mm.1 NM_177789.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00016784.mm.1 NM_177789.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00016784.mm.1 TC0X00002676.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0