This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00625351_g1
See other VSIG4 GE Assays ›
Gene Symbol:
Vsig4
Gene Name:
V-set and immunoglobulin domain containing 4
Gene Aliases:
A530061A11, BC025105, CRIg, Z39IG
Chromosome Location:
Chr.X: 96247201 - 96293518 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00625351_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Vsig4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177789.4 NP_808457.1 6 - 7 744 76
GenBank mRNA AK041014.1 - 6 - 7 745 76
BC025105.1 - 6 - 7 720 4162875 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00002676.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Vsig4

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s)

negative regulation of interleukin-2 production
negative regulation of T cell proliferation

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2012899 BC025105.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2012899 NM_177789.4 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2012899 AK041014.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 744855 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 8116 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 822699 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 212934 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 439643 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 41998 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 445175 AK006686.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 445175 AK133010.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 112129 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 261294 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 816497 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 141617 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 25414 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 313746 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 44762 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 165808 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 54203 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33389 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 121798 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 58617 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113325 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107256 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2012899 BC025105.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2012899 NM_177789.4 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2012899 AK041014.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32544650 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32544637 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32544643 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32544665 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32544658 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32544668 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32544652 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32544654 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32544657 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32544646 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32544635 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32544656 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32544648 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32544660 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32544662 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32544638 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32544655 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32544641 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32544534 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32544533 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32544627 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32544634 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32544631 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32544625 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32544640 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32544629 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09366692 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107256 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09366722 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09366707 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09366702 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113325 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03720915 - -
Whole Mouse Genome G4122A A_51_P128834 BC025105.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P128834 NM_177789.4 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P128834 AK041014.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131879 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107256 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09366701 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113325 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14930363 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14845830 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14845840 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14664790 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14664785 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14845832 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14664795 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14664791 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14546563 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544629 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544626 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544573 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544510 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544516 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544492 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544504 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544615 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544567 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544586 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544552 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544548 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544588 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544650 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544612 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544657 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544582 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544502 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544683 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544485 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544686 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544523 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544602 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544557 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544684 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544482 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544600 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544593 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544574 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544564 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544535 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544601 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544617 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544625 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544614 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544640 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544487 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544673 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544500 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544530 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544570 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544680 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544561 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544591 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544608 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544619 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544589 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544508 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544514 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544520 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544669 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544677 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544579 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544631 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544621 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544652 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544543 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544627 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544634 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544562 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544635 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544654 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544624 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544498 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544512 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544484 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544545 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544539 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544554 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544493 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544670 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544575 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544643 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544479 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544646 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544596 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544578 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544488 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544595 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544480 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544641 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544491 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544662 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544599 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544661 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544655 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544556 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544477 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544550 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544559 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544622 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544648 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544496 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544507 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544598 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544665 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544607 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544506 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544501 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544678 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544572 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544533 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544521 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544526 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544638 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32544541 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544665 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14635930 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14854831 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15223066 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544579 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544624 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544508 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544565 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544654 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544677 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544504 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544648 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544629 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544627 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544492 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544617 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544482 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544528 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14845833 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544530 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544621 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544634 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544641 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544655 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544669 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544561 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14845830 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544626 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544559 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15146016 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544541 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544652 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544576 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544671 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544502 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544638 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544485 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544480 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544520 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544496 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544556 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544498 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544516 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544514 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544562 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544578 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544477 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544668 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544564 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544619 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15036179 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544510 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544511 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544646 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544567 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14839027 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15204442 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14515355 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544673 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544500 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544640 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544479 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544524 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544650 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544535 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544493 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544507 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544533 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544611 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544523 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14844394 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544670 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544501 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544539 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544635 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14857512 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544660 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544661 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15204437 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544612 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544521 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544678 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544643 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14778977 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544657 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544555 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544631 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544491 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32544484 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10605851 - -
Affymetrix Mouse Gene 1.0 ST Array 10605849 - -
Affymetrix Mouse Gene 1.0 ST Array 10605852 - -
Affymetrix Mouse Gene 1.0 ST Array 10605853 - -
Affymetrix Mouse Gene 1.0 ST Array 10605850 - -
Affymetrix Mouse Gene 1.0 ST Array 10605854 - -
Affymetrix Mouse Gene 1.1 ST Array 10605848 - -
Mouse Genome 430 2.0 Array 1451651_PM_at - -
Mouse Genome 430 2.0 Array 1451651_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 278180_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 278180_at - -
Affymetrix Mouse Clariom S Array PSR0X00016780.mm.1 NM_177789.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00016780.mm.1 NM_177789.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00016780.mm.1 TC0X00002676.mm.1 MTA TC
Mouse Expression Set 430 1451651_PM_at - -
Mouse Expression Set 430 1451651_at - -
Murine Genome U74v2 Set 132172_at - -
Murine Genome U74v2 Set 132172_PM_at - -
Mouse Genome 430A 2.0 Array 1451651_at - -
Mouse Genome 430A 2.0 Array 1451651_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 678595 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 1580722 - -
Illumina mouse-6 v1.0 expression beadchip 1580722 - -
Illumina mouseRef-8 v1.0 expression beadchip 0001580722 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1241350 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1241350 - -
mouse-6 v1.1 (Illumina) scl54019.7.1_102-S - -
Illumina Sentrix mouse-6.v1 bead array 1580722 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl54019.7.1_102-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 1580722 - -
MouseRef-8 v2 Expression BeadChip ILMN_1241350 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1241350 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0