This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00626270_m1
See other RBM45 GE Assays ›
Gene Symbol:
Rbm45
Gene Name:
RNA binding motif protein 45
Gene Aliases:
Drb1, Drbp1, G430095G15Rik
Chromosome Location:
Chr.2: 76369984 - 76383768 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00626270_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbm45
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153405.2 NP_700454.1 8 - 9 1385 70
GenBank mRNA AB036992.1 - 8 - 9 1301 70
AK080420.1 - 8 - 9 1360 70
AK089975.1 - 8 - 9 1278 71
AK090084.1 - 8 - 9 1354 71
AK140861.1 - 8 - 9 1386 70
AY152391.1 - 8 - 9 1385 70
BC057890.1 - 8 - 9 1296 4979982 70
CB597326.1 - 2 - 3 230 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200001125.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rbm45

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
nervous system development
biological_process
cell differentiation
nucleotide binding
nucleic acid binding
RNA binding
poly(A) RNA binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21349792 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21349791 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21349788 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21349789 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21349790 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010906 AK080420.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010906 NM_153405.2 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 162901 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 396153 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 194562 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 312128 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 634183 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 825007 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 714339 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 253710 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 107237 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 55820 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 152626 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167088 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160970 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010906 AK080420.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010906 NM_153405.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P451374 BC057890.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P451374 AK089975.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P451374 AK090084.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P451374 AB036992.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21349805 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21349788 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21349809 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21349798 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21349794 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21349795 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21349796 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21349802 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21349806 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21349803 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21349800 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21349792 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21349801 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21349790 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160970 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00409248 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167088 - -
Whole Mouse Genome G4122A A_51_P451374 BC057890.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P451374 AK089975.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P451374 AK090084.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P451374 AB036992.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112992 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160970 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167088 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01768990 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21349790 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21349789 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21349792 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21349788 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21349791 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349913 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349864 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349885 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349921 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349748 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349764 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349909 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349774 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349886 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349814 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349895 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349773 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349796 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349862 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349846 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349805 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349858 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349924 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349751 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349882 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349938 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349843 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349770 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349899 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349828 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349822 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349839 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349866 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349747 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349936 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349900 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349792 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349887 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349849 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349816 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349845 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349901 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349853 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349750 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349897 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349801 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349884 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349923 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349875 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349798 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349888 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349755 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349818 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349761 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349889 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349860 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349941 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349769 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349780 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349870 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349783 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349829 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349892 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349871 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349926 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349819 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349812 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349903 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349759 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349915 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349914 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349912 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349811 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349746 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349806 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349893 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349867 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349865 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349904 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349917 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349841 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349775 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349844 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349760 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349827 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349856 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349905 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349757 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349809 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349790 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349833 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349836 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349779 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349800 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349902 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349919 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349825 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349762 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349823 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349799 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349855 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349803 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349880 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21349942 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349921 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01768988 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349838 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349776 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349829 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349814 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349909 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349822 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349779 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349889 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02032626 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349812 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349811 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349770 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349845 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349919 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02032634 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349805 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349803 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01936822 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349841 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01413399 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349880 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349888 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349800 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349915 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349787 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349844 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349887 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01329719 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349828 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349884 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349843 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349783 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01755698 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349873 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349836 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349825 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349799 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349839 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349874 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349780 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349819 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01329710 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01568152 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01329712 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349782 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349790 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349775 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349768 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01768980 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349911 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01936825 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349827 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349769 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349846 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349882 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349840 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349885 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349833 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349907 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01948491 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349796 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349914 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21349801 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10473053 - -
Affymetrix Mouse Gene 1.0 ST Array 10473049 - -
Affymetrix Mouse Gene 1.0 ST Array 10473046 - -
Affymetrix Mouse Gene 1.0 ST Array 10473050 - -
Affymetrix Mouse Gene 1.0 ST Array 10473048 - -
Affymetrix Mouse Gene 1.0 ST Array 10473051 - -
Affymetrix Mouse Gene 1.0 ST Array 10473057 - -
Affymetrix Mouse Gene 1.0 ST Array 10473054 - -
Affymetrix Mouse Gene 1.0 ST Array 10473047 - -
Affymetrix Mouse Gene 1.0 ST Array 10473055 - -
Affymetrix Mouse Gene 1.0 ST Array 10473052 - -
Affymetrix Mouse Gene 1.0 ST Array 10473056 - -
Affymetrix HT MG-430 PM Array Plate 1437904_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10473045 - -
Mouse Genome 430 2.0 Array 1437904_at - -
Mouse Genome 430 2.0 Array 1437904_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 241490_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 241490_at - -
Affymetrix Mouse Clariom S Array PSR0200009074.mm.1 NM_153405.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200009074.mm.1 NM_153405.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200009074.mm.1 TC0200001125.mm.1 MTA TC
Mouse Expression Set 430 1437904_at - -
Mouse Expression Set 430 1437904_PM_at - -
Murine Genome U74v2 Set 133930_PM_at - -
Murine Genome U74v2 Set 133930_at - -
Affymetrix HT Mouse Genome 430B Array 1437904_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1437904_at - -
Array Name Probe IDs Targets Target Type
Illumina mouseRef-8 v1.1 expression beadchip ILMN_3163340 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1247148 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_3163340 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1247148 - -
mouse-6 v1.1 (Illumina) GI_23510272-A - -
MouseRef-8 v2 Expression BeadChip ILMN_3163340 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_3163340 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) GI_23510272-A - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0