Assay Details
| Gene Symbol: Ndor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082476.2 | NP_001075945.1 | 2 - 3 | 268 | 69 | |
| NM_001252541.1 | NP_001239470.1 | 2 - 3 | 268 | 69 | ||
| NM_001252542.1 | NP_001239471.1 | 2 - 3 | 335 | 69 | ||
| GenBank mRNA | AK019617.1 | - | 2 - 3 | 265 | 69 | |
| AK143124.1 | - | 2 - 3 | 308 | 69 | ||
| AK163207.1 | - | 2 - 3 | 238 | 69 | ||
| AK169885.1 | - | 2 - 3 | 327 | 69 | ||
| BC049789.1 | - | 2 - 3 | 318 | 6331033 | 69 | |
| BQ771207.1 | - | 2 - 3 | 235 | 69 | ||
| CD354207.1 | - | 659 | 69 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001252542.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cell death
cellular response to menadione
oxidation-reduction process
NADPH-hemoprotein reductase activity
iron ion binding
FMN binding
oxidoreductase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen
epoxide carboxylase activity
flavin adenine dinucleotide binding
NADP binding
reduced coenzyme F420:NADP+ oxidoreductase activity
cell death
cellular response to menadione
oxidation-reduction process
NADPH-hemoprotein reductase activity
iron ion binding
FMN binding
oxidoreductase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen
epoxide carboxylase activity
flavin adenine dinucleotide binding
NADP binding
reduced coenzyme F420:NADP+ oxidoreductase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2082604 | BC049789.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2082604 | AK163207.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2082604 | NM_001082476.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2335695 | AK169885.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2335695 | AK019617.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2335695 | AK163207.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2335695 | NM_001082476.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 666528 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 224463 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 819300 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 614052 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 558826 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509470 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 45291 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2082604 | BC049789.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2082604 | AK163207.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2082604 | NM_001082476.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2335695 | AK169885.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2335695 | AK019617.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2335695 | AK163207.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2335695 | NM_001082476.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P310734 | AK169885.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P310734 | BC049789.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P310734 | AK019617.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P310734 | AK163207.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P310734 | NM_001082476.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P664800 | BC049789.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P664800 | AK143124.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P664800 | AK163207.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P664800 | NM_001082476.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P605129 | BQ771207.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P605129 | AK169885.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P605129 | BC049789.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P605129 | AK019617.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P605129 | AK143124.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01506207 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01506211 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01506225 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01506216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01801735 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01423248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02040354 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01506211 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02040395 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01801734 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02040396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01937214 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02040370 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02040412 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070851 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070852 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070860 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070868 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070892 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01423247 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02040348 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01506207 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02040403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02040378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01801731 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01887153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070835 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01801728 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070897 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02040362 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02040398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070847 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01937213 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070849 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02040350 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070871 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10480670 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480666 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480663 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480669 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480667 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480655 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480665 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480668 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480661 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480654 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480656 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480653 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480658 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480671 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480659 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480657 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480664 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480660 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480662 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428292_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435388_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10480652 | - | - |
| Mouse Genome 430 2.0 Array | 1435388_at | - | - |
| Mouse Genome 430 2.0 Array | 1428292_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428292_at | - | - |
| Mouse Genome 430 2.0 Array | 1435388_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78797_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78797_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200025851.mm.1 | NM_001082476.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200025851.mm.1 | NM_001082476.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200025851.mm.1 | NM_001252541.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200025851.mm.1 | NM_001252541.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200025851.mm.1 | NM_001252542.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200025851.mm.1 | NM_001252542.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200025851.mm.1 | TC0200003140.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200025853.mm.1 | TC0200003140.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200025850.mm.1 | NM_001082476.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200025850.mm.1 | NM_001082476.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200025850.mm.1 | NM_001252541.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200025850.mm.1 | NM_001252541.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200025850.mm.1 | NM_001252542.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200025850.mm.1 | NM_001252542.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200025850.mm.1 | TC0200003140.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200025855.mm.1 | NM_001252542.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200025855.mm.1 | NM_001252542.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200025855.mm.1 | TC0200003140.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428292_at | - | - |
| Mouse Expression Set 430 | 1435388_at | - | - |
| Mouse Expression Set 430 | 1435388_PM_at | - | - |
| Mouse Expression Set 430 | 1428292_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428292_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435388_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435388_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428292_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 588359 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 103190044 | - | - |
| MouseWG-6 v2 Expression BeadChip | 4200066 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 103190044 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1226703 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1226703 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1226135 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103190044 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103190044 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3146204 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3068081 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1226703 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1226703 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3068081 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3146204 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1226135 | - | - |

Ndor1
Genomic Map