This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00626705_m1
See other GPR107 GE Assays ›
Gene Symbol:
Gpr107
Gene Name:
G protein-coupled receptor 107
Gene Aliases:
9930033D15Rik, AI790205, C530034M11, mKIAA1624
Chromosome Location:
Chr.2: 31151997 - 31216567 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00626705_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr107
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178760.4 NP_848875.2 4 - 5 422 105
GenBank mRNA AK030433.1 - 4 - 5 418 105
AK034815.1 - 4 - 5 405 105
AK083025.1 - 4 - 5 416 105
AK088544.1 - 4 - 5 406 105
AK129407.1 - 4 - 5 317 105
AK142454.1 - 4 - 5 387 105
AK154530.1 - 4 - 5 418 105
BC092231.1 - 4 - 5 390 30607603 105
BC117900.1 - 4 - 5 400 40097045 105
BC117901.1 - 4 - 5 400 40097052 105

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpr107

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0