This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00626780_m1
See other SLC7A6 GE Assays ›
Gene Symbol:
Slc7a6
Gene Name:
solute carrier family 7 (cationic amino acid transporter, y+ system), member 6
Gene Aliases:
AI643885, LAT-2, LAT3
Chromosome Location:
Chr.8: 106168830 - 106198708 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00626780_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc7a6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178798.3 NP_848913.1 4 - 5 910 111
GenBank mRNA AK030762.1 - 5 - 6 712 111
AK049478.1 - 4 - 5 910 111
AK053899.1 - 4 - 5 919 111
AK129099.1 - 1 - 2 2306 111
AK157127.1 - 4 - 5 1049 111
AK164666.1 - 4 - 5 911 111
AK165629.1 - 4 - 5 927 111
AK165979.1 - 4 - 5 929 111
AK196234.1 - 341 111
AK197438.1 - 2 - 3 208 111
BC038404.1 - 3 - 4 844 4949962 111

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001245.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc7a6

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
transport
amino acid transport
L-alpha-amino acid transmembrane transport
molecular_function
amino acid transmembrane transporter activity
L-amino acid transmembrane transporter activity
antiporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800009987.mm.1 NM_178798.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800009987.mm.1 NM_178798.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800009987.mm.1 TC0800001245.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline